Learn More
Strem, An Ascensus Company CAS# 584-08-7. 50G. Potassium carbonate (99.997%-K) PURATREM. MFCD00011382
SDP
Catalog No. 501963601
Encompass
CAS# 584-08-7. 50G. Potassium carbonate (99.997%-K) PURATREM. MFCD00011382. Molecular Formula: K2CO3. Molecular Weight: 138.21. Color/Form: white pwdr. Stability: hygroscopic. Strem# 19-2600. www.strem.com/catalog/v3/19-2600/
Catalog No. 50-196-3601 Supplier Strem, An Ascensus Company Supplier No. 19260050G
CAS# 584-08-7. 50G. Potassium carbonate (99.997%-K) PURATREM. MFCD00011382. Molecular Formula: K2CO3. Molecular Weight: 138.21. Color/Form: white pwdr. Stability: hygroscopic. Strem# 19-2600. www.strem.com/catalog/v3/19-2600/