Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Potassium carbonate, 98%, extra pure, anhydrous, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 kg
5 kg
25 kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 891.0°C |
Quantity | 5 kg |
Linear Formula | K2CO3 |
Merck Index | 15,774 |
Solubility Information | Solubility in water: 1120g/L (20°C). Other solubilities: practically insoluble in ethanol, insoluble in acetone |
Formula Weight | 138.21 |
Percent Purity | 98% |
Grade | Extra Pure |
Loss on Drying | 2.0% max. |
Chemical Name or Material | Potassium carbonate |
Safety and Handling
Hazard Category | GHS Signal Word: Warning |
Hazard Statement | GHS H Statement May cause respiratory irritation. Causes skin irritation. Causes serious eye irritation. |
Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsing. If skin irri |
EINECSNumber | 209-529-3 |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction