Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Carbonate, (ACS Reagent Grade), MP Biomedicals

Catalog No. ICN15255894 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
5kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
ICN15255894 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15255894 Supplier MP Biomedicals, Inc Supplier No. 0215255894

Specifications

CAS 584-08-7
Quantity 5 kg
Molecular Formula CK2O3
MDL Number MFCD00011382
Synonym potassium carbonate, dipotassium carbonate, potash, potassium carbonate, anhydrous, carbonic acid, dipotassium salt, carbonate of potash, pearl ash, salt of tartar, kaliumcarbonat, potassiumcarbonate
InChI Key BWHMMNNQKKPAPP-UHFFFAOYSA-L
SMILES [K+].[K+].[O-]C([O-])=O
IUPAC Name dipotassium carbonate
Molecular Weight (g/mol) 138.21
PubChem CID 11430
Formula Weight 138.21
Grade ACS Reagent
Chemical Name or Material Potassium Carbonate, Anhydrous
Show More Show Less
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.