Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Carbonate, Anhydrous, Reagents

Catalog No. 01337121
Change view
Click to view available options
Quantity:
500 g
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-121 2.5 kg
01-337-120 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-337-121 Supplier Reagents Holdings Llc Supplier No. C22795002.5A5
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potassium Carbonate, Anhydrous, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 584-08-7
Molecular Formula CK2O3
Molecular Weight (g/mol) 138.21
MDL Number MFCD00011382
InChI Key BWHMMNNQKKPAPP-UHFFFAOYSA-L
Synonym Dipotassium Carbonate, Carbonate of Potash
IUPAC Name dipotassium carbonate
SMILES [K+].[K+].[O-]C([O-])=O

Specifications

Color Colorless/White
pH 11.6
Physical Form Solid
Quantity 2.5 kg
Grade Reagent
Packaging Poly Bottle
Specific Gravity 2.29
Chemical Name or Material Potassium Carbonate, Anhydrous
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.