Learn More
Potassium Carbonate Anhydrous (White Granular Powder), Fisher BioReagents
Shop All Fisherbrand ProductsDescription
Glass Bottle; 500g
Specifications
Specifications
| Chemical Name or Material | Potassium Carbonate Anhydrous |
| Melting Point | 891°C |
| Color | White |
| CAS | 584-08-7 |
| pH | 11.5 to 12 |
| Physical Form | Powder/Solid |
| Quantity | 500 g |
| Assay Percent Range | ≥99 % |
| Molecular Formula | CK2O3 |
| MDL Number | MFCD00011382 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.