Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Carbonate, puriss p.a, ACS, ≥99.0% (T), Solstice

Catalog No. 60047162 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
250 g
500 g
1 kg
2.5 kg
4 x 2.5 kg
Packaging:
Poly bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-047-162 500 g Poly bottle
60-047-159 250 g Poly bottle
60-047-157 1 kg Poly bottle
60-047-158 2.5 kg Poly bottle
60-047-161 4 x 2.5 kg Poly bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 60-047-162 Supplier Solstice Supplier No. 60109500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Puriss. p.a., ACS Reagent, anhydrous,≥99.0% (T)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 584-08-7
Molecular Formula CK2O3
Molecular Weight (g/mol) 138.21
MDL Number MFCD00011382
InChI Key BWHMMNNQKKPAPP-UHFFFAOYSA-L
Synonym potassium carbonate, dipotassium carbonate, potash, potassium carbonate, anhydrous, carbonic acid, dipotassium salt, carbonate of potash, pearl ash, salt of tartar, kaliumcarbonat, potassiumcarbonate
PubChem CID 11430
IUPAC Name dipotassium;carbonate
SMILES [K+].[K+].[O-]C([O-])=O

Specifications

Melting Point 891°C
Color Colorless or White
Physical Form Powder
Quantity 500 g
Linear Formula K2CO3
UN Number NONH for all modes of transport
Beilstein 4267587
Formula Weight 138.21g/mol
Percent Purity ≥99.0% (T)
Grade ACS
Packaging Poly bottle
Density 2.430g/cm3
Chemical Name or Material Potassium carbonate
Show More Show Less
Hazard Statement P261-P305 + P351 + P338
RTECSNumber 209-529-3
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.