Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Chlorate ACS AR Granular, Macron™

Catalog No. MK6834212 Shop All Avantor Products
Change view
Click to view available options
Glass; 4 x 2.5kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
MK-6834-212 Glass; 4 x 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MK-6834-212 Supplier Avantor Supplier No. 683406

missing translation for 'validMainframeMsg'

Specifications

CAS 3811-04-9
Physical Form Granules
Quantity 4 x 2.5 kg
Molecular Formula ClKO3
Synonym potassium chlorate, fekabit, potcrate, kaliumchlorat, anforstan, berthollet salt, salt of tarter, berthollet's salt, chlorate of potash, chloric acid, potassium salt
InChI Key VKJKEPKFPUWCAS-UHFFFAOYSA-M
SMILES [O-]Cl(=O)=O.[K+]
IUPAC Name potassium;chlorate
Molecular Weight (g/mol) 122.55
PubChem CID 6426889
Formula Weight 122.55
Grade ACS
Packaging Glass
Chemical Name or Material Potassium Chlorate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.