Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Chlorate, Crystal, ACS, 99%, Spectrum™ Chemical
SDP

Catalog No. 18612529 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-612-529 25 g Amber Glass Bottle
18-612-530 125 g Amber Glass Bottle
18-612-531 500 g Amber Glass Bottle
18-612-532 2.5 kg Amber Glass Bottle
18-612-533 12 kg Poly Pail
18-612-534 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-612-529 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P124525GM
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3811-04-9
Molecular Formula ClKO3
Molecular Weight (g/mol) 122.55
InChI Key VKJKEPKFPUWCAS-UHFFFAOYSA-M
IUPAC Name potassium chlorate
SMILES [K+].[O-][Cl](=O)=O

Specifications

CAS Max % 1
Quantity 25 g
Percent Purity 99%
Grade ACS
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.