Learn More
Biotang Inc Potassium Chloride 1M.1 M Potassium chloride (KCl) solution. Prepared in 18.2 megohms water and autoclaved
Catalog No. NC0790849 Shop All BIOTANG Inc Products
Encompass_Preferred
Potassium Chloride 1M.1 M Potassium chloride (KCl) solution. Prepared in 18.2 megohms water and autoclaved
Catalog No. NC0790849 Supplier Biotang Inc Supplier No. MT250
Potassium Chloride 1M.1 M Potassium chloride (KCl) solution. Prepared in 18.2 megohms water and autoclaved