Learn More
Biotang Inc POTASSIUM CHLORIDE 1M 50 ML, MT-250, 1 M Potassium chloride (KCl) solution. Prepared in 18.2 megohms water and autoclaved at 15 psi and 121 degree Celsius for 30 min on liquid cycle. Shipped at room temperature. Store at room temperature.
Catalog No. 50152273 Shop All BIOTANG Inc Products
Encompass_Preferred
POTASSIUM CHLORIDE 1M 50 ML, MT-250, 1 M Potassium chloride (KCl) solution. Prepared in 18.2 megohms water and autoclaved at 15 psi and 121 degree Celsius for 30 min on liquid cycle. Shipped at room temperature. Store at room temperature.
Catalog No. 50-152-273 Supplier Biotang Inc Supplier No. BTBB820
POTASSIUM CHLORIDE 1M 50 ML, MT-250, 1 M Potassium chloride (KCl) solution. Prepared in 18.2 megohms water and autoclaved at 15 psi and 121 degree Celsius for 30 min on liquid cycle. Shipped at room temperature. Store at room temperature.