Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium chloride, 1M aq. soln., RNAse free

Catalog No. AAJ62422AK
Change view
Click to view available options
Quantity:
100 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62422AK 250 mL
AAJ62422AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ62422AK Supplier Thermo Scientific Chemicals Supplier No. J62422AK
Add to Cart

CAS: 7447-40-7 | ClK | 74.55 g/mol

It may be used for the preparation of phosphate buffered saline, and for the extraction and solubilization of proteins. This solution is used in a variety of biological applications, including DNA and RNA isolation techniques. Also used to establish a constant ionic strength in a sample solution. pH: 5.0-8.0 (25°C, 1 M in H2O)

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It may be used for the preparation of phosphate buffered saline, and for the extraction and solubilization of proteins. This solution is used in a variety of biological applications, including DNA and RNA isolation techniques. Also used to establish a constant ionic strength in a sample solution. pH: 5.0-8.0 (25°C, 1 M in H2O)

Solubility
Not miscible in water.

Notes
Store at room temperature. Ensure good ventilation.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7447-40-7
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
Synonym potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
PubChem CID 4873
ChEBI CHEBI:32588
SMILES [Cl-].[K+]

Specifications

Physical Form Liquid
Quantity 250 mL
Linear Formula KCl
Solubility Information Not miscible in water.
IUPAC Name potassium chloride
Formula Weight 74.55
Concentration or Composition (by Analyte or Components) 1M aq. soln.
Chemical Name or Material Potassium chloride, RNAse free
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.