Learn More
Potassium Chloride, 3 Molar Electrode Filling Solution, Reagents
Specifications
Specifications
| Chemical Name or Material | Potassium Chloride, 3 Molar Electrode Filling Solution |
| Boiling Point | 103°C |
| Color | Colorless |
| CAS | 7732-18-5, 7447-40-7 |
| pH | 5.4 to 8.6 |
| Physical Form | Liquid |
| Quantity | 500 mL |
| Molecular Formula | KCl/H2O |
| Synonym | Muriate of Potash |
| Grade | Reagent |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.