Learn More
Research Products International Corp Potassium Chloride, 500 Grams
SDP
Supplier:  Research Products International Corp P41000500.0
Encompass_Preferred
Potassium chloride (KCl) is a white, water-soluble salt (74.55 g/mol) used in life science labs to maintain ionic strength and osmotic balance. It supports cell culture, electrophysiology, and buffer preparation, helping regulate ion gradients in biochemical assays, protein purification, and molecular biology workflows requiring controlled potassium levels.
Catalog No. 50-488-925