Learn More
Medchemexpress LLC Potassium chloride | 7447-40-7 | MFCD00011360 | 99.5% | 74.55 g/mol | KCl | 25g
SDP
Catalog No. 5000268984
Encompass_Preferred
Potassium chloride for molecular biology is potassium chloride that can be used in molecular biology Potassium chloride for molecular biology affects the stability of biological membranes by disrupting the electrostatic interactions between proteins and lipids Potassium chloride for molecular biology affects the solubility of myofibrillar proteins and the integrity of mitochondria Potassium chloride for molecular biology is commonly used in homogenization buffers and protein extraction procedures[1][2][3][4][5][6][7]
Catalog No. 50-002-68984 Supplier Medchemexpress LLC Supplier No. HYY0537B25G
Potassium chloride for molecular biology is potassium chloride that can be used in molecular biology Potassium chloride for molecular biology affects the stability of biological membranes by disrupting the electrostatic interactions between proteins and lipids Potassium chloride for molecular biology affects the solubility of myofibrillar proteins and the integrity of mitochondria Potassium chloride for molecular biology is commonly used in homogenization buffers and protein extraction procedures[1][2][3][4][5][6][7]