Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium chloride, 99+%, for analysis

Catalog No. AC196770050
Change view
Click to view available options
Quantity:
1 kg
5 kg
25 kg
Packaging:
Fibre drum
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC196770050 5 kg Plastic bottle
AC196770010 1 kg Plastic bottle
AC196770250 25 kg Fibre drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC196770050 Supplier Thermo Scientific Chemicals Supplier No. 196770050
Only null left
Add to Cart

CAS: 7447-40-7 | ClK | 74.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer for analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7447-40-7
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
Synonym potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
PubChem CID 4873
ChEBI CHEBI:32588
SMILES [Cl-].[K+]

Specifications

Boiling Point 1420.0°C
Melting Point 770.0°C
Color Colorless
Physical Form Liquid
Quantity 5 kg
Assay Percent Range 99.0 to 100.5% (on dried substance) (Argentometry)
Linear Formula KCl
Fieser 06,48
Merck Index 15,7742
Solubility Information Solubility in water: 340g/L (20°C). Other solubilities: slightly soluble in alcohol,insoluble in ether and acetone
IUPAC Name potassium;chloride
Formula Weight 74.54
Percent Purity ≥99%
Grade Analytical
Loss on Drying 0.5 % max. (1 g, 105°C, 3 hrs)
Packaging Plastic bottle
Chemical Name or Material Potassium chloride, p.a.
Show More Show Less
Hazard Category HYGROSCOPIC
EINECSNumber 231-211-8
RTECSNumber TS8050000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.