Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Chloride, >99%, MP Biomedicals™

Catalog No. 021519442 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
250 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02151944-2 250 g
02151944-5 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02151944-2 Supplier MP Biomedicals, Inc Supplier No. 02151944.2
Add to Cart
Add to Cart

Potassium Chloride, >99% is used for the extraction and solubilization of proteins.

  • Potassium chloride is a salt commonly used in the preparation of phosphate buffered saline and for the extraction and solubilization of proteins.
  • It is a good potassium source.
  • It is also used in the hypotonic KCl-propidium iodide protocol for univariate analysis of metaphase chromosomes.
  • It is used for the dissociation of eukaryotic ribosomes into subunits by high concentrations of potassium (0.3-1.0 M).
  • Potassium chloride is used in the preparation of mitochondria.
  • It also provides an ionic osmotic support for those tissues that assume a gelatinous consistency upon homogenization.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7447-40-7
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
IUPAC Name potassium chloride
SMILES [Cl-].[K+]

Specifications

Boiling Point 2732°F (Sublimes)
Melting Point 1418°F
pH ∼7
Physical Form Crystalline Powder
Quantity 250 g
Solubility Information >100 mg/mL at 68°F
Percent Purity 99%
Density 1.984 g/cm3
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.