Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Chloride, ACS Reagent, Reagents

Catalog No. 01337863
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-863 125 g
01-337-862 12 kg
01-337-861 2.5 kg
01-337-860 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01-337-863 Supplier Reagents Holdings Llc Supplier No. C1280500125D2
 Restricted Item
Add to Cart
Add to Cart

Potassium Chloride, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7447-40-7
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
Synonym Muriate of Potash
IUPAC Name potassium chloride
SMILES [Cl-].[K+]

Specifications

Boiling Point 1413°C
Melting Point 771°C
Color Colorless/White
Physical Form Solid
Quantity 125 g
Grade ACS
Packaging Glass Bottle
Specific Gravity 1.984
Chemical Name or Material Potassium Chloride, ACS Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.