Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Chloride Conductivity Standard, 250 μS/cm at 25°C, Ricca Chemical
SDP

Catalog No. R58873004A Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
Poly
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R58873004A 4 L Natural Poly Bottle
R5887300500 500 mL Natural Poly Bottle
58-873-0032 1 L Poly
R588730010F 10 L Cubitainer
R588730020F 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. R58873004A Supplier Ricca Chemical Company Supplier No. R58873004A
Add to Cart
Add to Cart

Potassium Chloride Conductivity Standard is tested in ISO 17025 accredited laboratories, providing the utmost quality in calibration.

  • Tested in our ISO 17025 Accredited Laboratories.
  • Manufactured in cGMP compliant, FDA registered facilities.
  • Certified Traceable to NIST Standard Reference Material when available.
  • Many C of As and labels contain TDS and conductance information.
  • Tightest Specifications in the industry, lowest lot-to-lot variability.

Chemical Identifiers

CAS 7447-40-7 , 7732-18-5
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
PubChem CID 4873
ChEBI CHEBI:32588
IUPAC Name potassium chloride
SMILES [Cl-].[K+]

Specifications

Packaging Natural Poly Bottle
Odor Odorless
Assay Percent Range 250 ± 1 μS/cm at 25°C
Physical Form Liquid
Color Colorless
Boiling Point 100°C
Solubility Infinitely soluble in water
Solution Type Conductivity Standard
Type Conductivity Standard
Solvent or Matrix Water
Quantity 4 L
Chemical Name or Material Potassium Chloride Conductivity Standard, 250 μS/cm (250 μmho/cm) at 25°C
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.