Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Chloride Conductivity Standard, 700 μS/cm at 25°C, Ricca Chemical
SDP

Catalog No. 5887716 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
58877-16 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 58877-16 Supplier Ricca Chemical Company Supplier No. 5887.716
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7447-40-7 , 7732-18-5
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
PubChem CID 4873
ChEBI CHEBI:32588
IUPAC Name potassium chloride
SMILES [Cl-].[K+]

Specifications

Packaging Natural Poly Bottle
CAS Min % 98.0, 0.03
CAS Max % 100.0, 0.03
Physical Form Liquid
Color Colorless
pH Approximately 7
Melting Point 0.0°C
Boiling Point 100°C
For Use With (Equipment) Conductivity Meters and Probes
Solubility Miscible with water
Solution Type Conductivity Standard
Type Conductivity Standard
Quantity 500 mL
Grade Laboratory
Concentration or Composition (by Analyte or Components) 700 μS/cm at 25°C
Chemical Name or Material Potassium Chloride, Conductivity Standard
Show More Show Less
ShelfLife 12 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.