Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Chloride, Crystal, Ultrapure Bioreagent, J.T. Baker™

Catalog No. 02003743 Shop All Avantor Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02-003-743 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-003-743 Supplier Avantor Supplier No. 400101
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

    For Molecular Biology Applications

Chemical Identifiers

CAS 7447-40-7
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
Synonym potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
PubChem CID 4873
ChEBI CHEBI:32588
SMILES [Cl-].[K+]

Specifications

Physical Form Crystals
Quantity 500 g
IUPAC Name potassium;chloride
Formula Weight 74.55
Grade Ultrapure Bioreagent
Packaging Poly Bottle
Specific Gravity 1L = 1.99kg
Chemical Name or Material Potassium Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.