Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Chloride, Crystal, USP, 99-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18605500 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
45 kg
113.5 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-605-500 500 g Poly Bottle
18-605-502 2.5 kg Poly Bottle
18-605-501 6 x 500 g Poly Bottle
18-605-503 4 x 2.5 kg Poly Bottle
18-605-504 12 kg Poly Pail
18-605-505 45 kg Fiber Drum
18-605-506 113.5 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18-605-500 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. PO165500GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

PO165, 7447-40-7, KCl

Spectrum™ Chemical Potassium Chloride, Crystal, USP is used to treat hypokalemia. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7447-40-7
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
IUPAC Name potassium chloride
SMILES [Cl-].[K+]

Specifications

CAS Max % 1
Quantity 500 g
Percent Purity 99 to 100.5%
Grade USP
Loss on Drying 0.01
Packaging Poly Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.