Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Chloride, For Analysis, EMSURE™, (≤ 0.005% Br), MilliporeSigma™

Catalog No. M1049339025 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 kg
Packaging:
Fiber Carton
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1049339025 25 kg Fiber Carton
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1049339025 Supplier MilliporeSigma Supplier No. 1049339025
Add to Cart
Add to Cart
Request a Quote

Chemical Identifiers

CAS 7447-40-7
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
Synonym potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
PubChem CID 4873
ChEBI CHEBI:32588
IUPAC Name potassium chloride
SMILES [Cl-].[K+]

Specifications

Boiling Point 1413°C (1013hPa)
Melting Point 773°C
Color White
pH 5.5 to 8.0 (50g/L, H2O, 25°C)
Physical Form Solid
Quantity 25 kg
Solubility Information 347g/L
Formula Weight 74.55
Percent Purity 99.5 to 100.5%
Grade ACS
Packaging Fiber Carton
Density 1.98 g/3 (20 °C)
Chemical Name or Material Potassium Chloride
Show More Show Less
EINECSNumber 231-211-8
RTECSNumber TS8050000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.