Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Chloride, Granular, BiotechGrade, 99-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18605507 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-605-507 500 g Poly Bottle
18-605-508 2.5 kg Poly Bottle
18-605-509 12 kg Poly Pail
18-605-510 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-605-507 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P1251500GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

P1251, 7447-40-7, KCl

Spectrum™ Chemical Potassium Chloride, Granular, BiotechGrade is commonly used as a laboratory reagent as a standard. It may be used as a calibration standard ionic solution for measuring electrical conductivity because potassium chloride solutions, when prepared carefully, produce repeatable measurable properties. Potassium chloride is also a good source of ionic chloride, which should precipitate insoluble chloride salts upon addition to a solution of an appropriate metal ion.

Chemical Identifiers

CAS 7447-40-7
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
IUPAC Name potassium chloride
SMILES [Cl-].[K+]

Specifications

CAS Max % 1
Quantity 500 g
Percent Purity 99 to 100.5%
Grade Biotech
Loss on Drying 0.01
Odor Odorless
Packaging Poly Bottle
Appearance Clear
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.