Learn More
Crescent Chemical Co Inc POTASSIUM CHLORIDE MOL BIO GRD
SDP
Catalog No. 50248454
Encompass_Preferred
Potassium chloride molecular biology grade DNase/RNase not detected. Size - 500G Storage Conditions - +15 °C TO +30 °C Catalog Number - 39768.01 CAS 7447-40-7
Catalog No. 50-248-454 Supplier Crescent Chemical Co Inc Supplier No. SE39768.01
Potassium chloride molecular biology grade DNase/RNase not detected. Size - 500G Storage Conditions - +15 °C TO +30 °C Catalog Number - 39768.01 CAS 7447-40-7