Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Chloride, Purified, Reagents

Catalog No. 01337116
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-116 12 kg
01-337-115 2.5 kg
01-337-113 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-337-116 Supplier Reagents Holdings Llc Supplier No. C228050012KG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potassium Chloride, Purified, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7447-40-7
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
Synonym Muriate of Potash
IUPAC Name potassium chloride
SMILES [Cl-].[K+]

Specifications

Boiling Point 1413°C
Melting Point 771°C
Color Colorless/White
Physical Form Solid
Quantity 12 kg
Percent Purity 98.92%
Grade Pure
Packaging Poly Pail
Specific Gravity 1.984
Chemical Name or Material Potassium Chloride, Purified
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.