Learn More
Sigma Aldrich Fine Chemicals Biosciences Potassium chloride puriss.1KG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 126361KG
Encompass
Potassium chloride an alkali metal halide is a colorless crystalline salt. Its solubility in water is directly proportional to increase in temperature. It can be prepared from potassium hydroxide via neutralization reaction with HCl. Calorimetric estimation of the enthalpy of SOLUTIONon of potassium chloride in water has been described.
Catalog No. 50-289-7232