Learn More
Sigma Aldrich Fine Chemicals Biosciences Potassium chloride puriss.250G
Catalog No. 502898804
Encompass
Potassium chloride an alkali metal halide is a colorless crystalline salt. Its solubility in water is directly proportional to increase in temperature. It can be prepared from potassium hydroxide via neutralization reaction with HCl. Calorimetric estimation of the enthalpy of SOLUTIONon of potassium chloride in water has been described.
Catalog No. 50-289-8804 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 12636250G
Potassium chloride an alkali metal halide is a colorless crystalline salt. Its solubility in water is directly proportional to increase in temperature. It can be prepared from potassium hydroxide via neutralization reaction with HCl. Calorimetric estimation of the enthalpy of SOLUTIONon of potassium chloride in water has been described.