Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Chloride, puriss. p.a., Solstice

Catalog No. 6002329 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
4 x 5 kg
6 x 1 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-023-29 500 g
60-023-28 1 kg
60-023-30 5 kg
60-045-928 6 x 1 kg
60-045-927 4 x 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 60-023-29 Supplier Solstice Supplier No. 31248500G
Add to Cart
Add to Cart

Puriss. p.a., Reag. ISO, Reag. Ph. Eur., 99.5-100.5%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7447-40-7
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
Synonym potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
PubChem CID 4873
ChEBI CHEBI:32588
SMILES [Cl-].[K+]

Specifications

Quantity 500 g
Linear Formula KCl
UN Number NONH for all modes of transport
IUPAC Name potassium chloride
Formula Weight 74.55g/mol
Percent Purity 99.5 to 100.5%
Grade puriss. p.a.
Density 1.98g/mL at 25°C
Chemical Name or Material Potassium Chloride, Reag. Ph. Eur. reag. ISO
EINECSNumber 231-211-8
RTECSNumber TS8050000
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.