Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Chloride Solution, Conductance Standard B acc. to ISO 7888, 0.01 M KCl, MilliporeSigma™ Supelco™

Catalog No. 111015416 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5416 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5416 Supplier MilliporeSigma Supelco Supplier No. 60136250ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 7447-40-7
Concentration 0.01 M KCl
Linear Formula KCl
MDL Number MFCD00011360
Quantity 250 mL
Formula Weight 74.55
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.