Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium chloride solution, MilliporeSigma™

Catalog No. M1048170250 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 mL
Packaging:
Plastic Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1048170250 250 mL Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1048170250 Supplier MilliporeSigma Supplier No. 1.04817.0250
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7447-40-7
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
Synonym potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
PubChem CID 4873
ChEBI CHEBI:32588
IUPAC Name potassium chloride
SMILES [Cl-].[K+]

Specifications

Quantity 250 mL
Solubility Information Soluble in water at 20°C
Odor Odorless
Packaging Plastic Bottle
Density 1.13g/cm3 at 20°C
Chemical Name or Material Potassium chloride solution
Recommended Storage Store at 15° to 25°C (59° to 77°F), Tightly closed.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.