Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Chloride Solution, Solstice

Catalog No. 60047165 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-047-165 500 mL Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-047-165 Supplier Solstice Supplier No. 60131500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

conductivity 111.8 mS/cm at 25 °C, conductance standard

Solstice's high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use
  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7447-40-7
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
Synonym potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
PubChem CID 4873
ChEBI CHEBI:32588
IUPAC Name potassium;chloride
SMILES [Cl-].[K+]

Specifications

Color Colorless
Physical Form Liquid
Quantity 500 mL
Linear Formula KCl
UN Number NONH for all modes of transport
Beilstein 1711999
Solubility Information Completely soluble in water
Formula Weight 74.55g/mol
Packaging Plastic bottle
Density 1.04g/mL at 20°C
Chemical Name or Material Potassium chloride solution, Conductivity 111.8mS/cm at 25°C, conductance standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.