Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium chloride, ultra dry, 99.998% (metals basis)

Catalog No. AA1368209
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1368209 10 g
AA1368218 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1368209 Supplier Thermo Scientific Chemicals Supplier No. 01368209
Only null left
Add to Cart

CAS: 7447-40-7 | ClK | 74.55 g/mol

The main use of potassium chloride is in the manufacture of fertilizers. It finds use in medicine as an electrolyte replenisher and to treat hypokalemia (a condition in which the level of potassium will be low in blood) with its pharmaceutical preparation. Furthermore, it finds use as a flux agent in welding and metal casting. It is useful in water softener units, certain de-icing products and calibration of the radiation monitoring equipment. It finds use as a scotophor in dark-trace CRTs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7447-40-7
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
Synonym potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
PubChem CID 4873
ChEBI CHEBI:32588
SMILES [Cl-].[K+]

Specifications

Boiling Point 1500°C (sublimation)
Melting Point 773°C
Color White
Physical Form Beads
Quantity 10 g
Assay Percent Range 99.998% (metals basis)
Solubility Information Freely soluble in water. Slightly soluble in alcohol. Insoluble in ether.
IUPAC Name potassium chloride
Formula Weight 74.55
Odor Odorless
Refractive Index 1.49
Sensitivity Hygroscopic
Appearance White beads
Density 1.984 g/cm3
Chemical Name or Material Potassium chloride
Show More Show Less
Hazard Category H303: May be harmful if swallowed.
EINECSNumber 231-211-8
RTECSNumber TS8050000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.