Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Chromate ACS AR Crystal, Macron Fine Chemicals™

Catalog No. MK6870500 Shop All Avantor Products
Change view
Click to view available options
Quantity:
125 g
12 x 500 g
Packaging:
POLYSTORMOR
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MK-6870-500 12 x 500 g POLYSTORMOR
MK-6870-125 125 g POLYSTORMOR
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MK-6870-500 Supplier Avantor Supplier No. 687004

Chemical Identifiers

CAS 7789-00-6
Molecular Formula CrK2O4
Molecular Weight (g/mol) 194.19
MDL Number MFCD00011368
InChI Key XMXNVYPJWBTAHN-UHFFFAOYSA-N
Synonym potassium chromate, bipotassium chromate, potassium chromate vi, dipotassium monochromate, dipotassium chromate, tarapacaite, chromate of potash, chromate of potass, neutral potassium chromate, caswell no. 687
PubChem CID 24597
ChEBI CHEBI:75249
IUPAC Name dipotassium dioxochromiumbis(olate)
SMILES [K+].[K+].[O-][Cr]([O-])(=O)=O

Specifications

Physical Form Crystalline
Quantity 12 x 500 g
Formula Weight 194.19
Grade ACS
Packaging POLYSTORMOR
Chemical Name or Material Potassium Chromate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.