Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Chromate, ACS Reagent, Reagents

Catalog No. 01337859
Change view
Click to view available options
Quantity:
125 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-859 125 g
01-337-858 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-337-859 Supplier Reagents Holdings Llc Supplier No. C1281000125D2
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potassium Chromate, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7789-00-6
Molecular Formula CrK2O4
Molecular Weight (g/mol) 194.19
InChI Key XMXNVYPJWBTAHN-UHFFFAOYSA-N
Synonym Dipotassium monochromate
IUPAC Name dipotassium dioxochromiumbis(olate)
SMILES [K+].[K+].[O-][Cr]([O-])(=O)=O

Specifications

Boiling Point 1000°C
Color Yellow
Physical Form Solid
Quantity 125 g
Percent Purity 99.3%
Grade ACS
Packaging Glass Bottle
Specific Gravity 2.73
Chemical Name or Material Potassium Chromate, ACS Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.