Learn More
Hach Company Potassium Chromate Solution, 50 g/L, 500mL
Supplier:  Hach Company 16649H
Encompass
Indicator for chloride determination by the argentometric titration method
Catalog No. 50-199-7380