Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Chromate TS, 10% w/v Chloride Free, Indicator for Argentometric Titrations, Reagents

Catalog No. 01334349
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-334-349 500 mL
01-334-350 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-334-349 Supplier Reagents Holdings Llc Supplier No. CP114100500A
Add to Cart
Add to Cart

Potassium Chromate TS, 10% w/v Chloride Free, Indicator for Argentometric Titrations, Meets USP specifications, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Potassium Chromate TS, 10% w/v Chloride Free, Indicator for Argentometric Titrations
Boiling Point 102°C
Color Yellow
CAS 7732-18-5, 7789-00-6
Physical Form Liquid
Quantity 500 mL
Molecular Formula K2CrO4/H2O
Synonym Dipotassium monochromate
Grade USP
Packaging Poly Bottle
Specific Gravity 1.0653
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.