Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium cubes, 99.5% (metals basis), Thermo Scientific™

Catalog No. AA4668713
Change view
Click to view available options
Quantity:
20 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4668713 20 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4668713 Supplier Thermo Scientific Chemicals Supplier No. 4668713

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Packaged in mineral oil
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9-7-7440
Molecular Formula K
Molecular Weight (g/mol) 39.10
MDL Number MFCD00133776
InChI Key ZLMJMSJWJFRBEC-UHFFFAOYSA-N
Synonym kalium, potassium, metal, potassium, liquid alloy, unii-rwp5ga015d, rwp5ga015d, hsdb 698, monopotassium, potasio, mono-potassium, atom
PubChem CID 5462222
ChEBI CHEBI:26216
IUPAC Name potassium
SMILES [K]

Specifications

Quantity 20 g
Odor Odorless
Weight ≈20g/cube
Form Cube
UN Number UN2257
Percent Purity 99.5%
Assay (metals basis)
Chemical Name or Material Potassium cubes
Hazard Category H260-H314-H335
Hazard Statement GHS H Statement
H260-H314-H318
In contact with water releases flammable gases which may ignite spontaneously.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P223-P231+P232-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P335+P334-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: POTASSIUM
EINECSNumber 231-119-8
RTECSNumber TS6460000
TSCA Yes
storageNote1
  • Packaged in mineral oil
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.