Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Cyanide, For Analysis, EMSURE™, MilliporeSigma™

Catalog No. M1049670250 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 g
Packaging:
Plastic Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1049670250 250 g Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1049670250 Supplier MilliporeSigma Supplier No. 1.04967.0250
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 151-50-8
Molecular Formula CKN
Molecular Weight (g/mol) 65.116
InChI Key NNFCIKHAZHQZJG-UHFFFAOYSA-N
Synonym potassium cyanide, potassium cyanide k cn, cyanide of potassium, caswell no. 688a, zyankali, kalium cyanid german, unii-mqd255m2zo, rcra waste number p098, cyanure de potassium french
PubChem CID 9032
IUPAC Name potassium;cyanide
SMILES [C-]#N.[K+]

Specifications

Boiling Point 1625°C (1013hPa)
Melting Point 634°C
Color White
pH 11 to 12
Physical Form Solid
Quantity 250 g
Solubility Information 716g/L
Formula Weight 65.12
Percent Purity >97.0%
Grade ACS
Packaging Plastic Bottle
Density 1.55 g/3 (20 °C)
Show More Show Less
EINECSNumber 205-792-3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.