Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Cyanide, puriss. p.a. ACS Reagent, Solstice

Catalog No. 6001207 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
250 g
1 kg
25 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-012-07 25 kg
60-012-04 100 g
60-012-06 250 g
60-012-05 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 60-012-07 Supplier Solstice Supplier No. 3125225KG
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Puriss. p.a., ACS Reagent, Reag. Ph. Eur.,≥97.0%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 151-50-8
Molecular Formula CKN
Molecular Weight (g/mol) 65.116
MDL Number MFCD00011397
InChI Key NNFCIKHAZHQZJG-UHFFFAOYSA-N
Synonym potassium cyanide, potassium cyanide k cn, cyanide of potassium, caswell no. 688a, zyankali, kalium cyanid german, unii-mqd255m2zo, rcra waste number p098, cyanure de potassium french
PubChem CID 9032
IUPAC Name potassium;cyanide
SMILES [C-]#N.[K+]

Specifications

Physical Form Solid
Quantity 25 kg
Linear Formula KCN
UN Number 1680
Beilstein 3593645
Formula Weight 65.12g/mol
Percent Purity ≥97%
Grade Not Available
Chemical Name or Material Potassium Cyanide, Reag. Ph. Eur.
Hazard Statement P260-P264-P273-P280-P284-P301 + P310
Precautionary Statement H290-H300 + H310 + H330-H370-H372-H410
EINECSNumber 205-792-3
RTECSNumber TS8750000
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.