Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium cyclohexanebutyrate, K 16.6-18.8%, Thermo Scientific™

Catalog No. AAA1783914
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1783914 25 g
AAA1783903 1 g
AAA1783906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1783914 Supplier Thermo Scientific Chemicals Supplier No. A1783914

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62638-03-3
Molecular Formula C10H17KO2
Molecular Weight (g/mol) 208.34
MDL Number MFCD00013051
InChI Key LBYNWSSEILNGLN-UHFFFAOYSA-M
Synonym potassium cyclohexanebutyrate, potassium 4-cyclohexylbutanoate, potassium 4-cyclohexylbutyrate, cyclohexanebutanoic acid, potassium salt, cyclohexanebutanoic acid, potassium salt 1:1, acmc-20ajuw, cyclohexanebutyric acid potassium salt, cyclohexanebutanoic acid potassium salt, 4-cyclohexylbutanoic acid, potassium salt
PubChem CID 23692296
IUPAC Name potassium;4-cyclohexylbutanoate
SMILES [K+].[O-]C(=O)CCCC1CCCCC1

Specifications

Quantity 25 g
Sensitivity Hygroscopic
Formula Weight 208.33
Assay K 16.6 to 18.8%
Chemical Name or Material Potassium cyclohexanebutyrate
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 263-666-3
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.