Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Potassium dehydroandrographolide succinate | 76958-99-1 | 99.3% | 570.67 g/mol | C28H35KO10 | 10 MG
SDP

Supplier:  Medchemexpress LLC HYN0677A10MG

Encompass_Preferred

Potassium dehydroandrographolide succinate is a research chemical derived from the herbal medicine Andrographis paniculata. It is studied for immunostimulatory, anti-infective, and anti-inflammatory activities and is used in preclinical research on viral respiratory infections.

  • Purity: 99.3%.
  • Molecular formula: C28H35KO10; molecular weight: 570.67 g/mol.
  • CAS number: 76958-99-1.
  • Typical package sizes: 5 mg, 10 mg, 20 mg.
  • Supplied as a solid for in vitro and in vivo research applications.

Catalog No. 50-002-95509


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.