Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Dichromate, 0.0250 N (N/40), Ricca Chemical
SDP

Catalog No. R6050000100 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R6050000100 100 mL Natural Poly Bottle
6050-16 500 mL Natural Poly Bottle
6050-32 1 L Natural Poly Bottle
6050-1 4 L Natural Poly Bottle
R605000020F 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. R6050000100 Supplier Ricca Chemical Company Supplier No. 60504
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For ASTM/EPA Testing

Chemical Identifiers

Molecular Formula Cr2K2O7
Molecular Weight (g/mol) 294.182
InChI Key KMUONIBRACKNSN-UHFFFAOYSA-N
PubChem CID 24502
ChEBI CHEBI:53444
IUPAC Name dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
SMILES [O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+]

Specifications

Color Colorless
CAS 7778-50-9, 7732-18-5
Assay Percent Range 0.02500 ± 0.00001 N at 20°C
Solubility Information Solubility in Water: Infinite
Grade Laboratory
Odor Odorless
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
pH ∼4
Quantity 100 mL
Physical Form Liquid
Chemical Name or Material Potassium Dichromate, 0.0250N (N/40)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.