Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Dichromate, 0.200 N (N/5), Ricca Chemical
SDP

Catalog No. 606516 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
606516 500 mL
6065-32 1 L
6065-1 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 606516 Supplier Ricca Chemical Company Supplier No. 606516
Add to Cart
Add to Cart
This item is not returnable. View return policy

Chemical Identifiers

Molecular Formula Cr2K2O7
InChI Key KMUONIBRACKNSN-UHFFFAOYSA-N
PubChem CID 24502
ChEBI CHEBI:53444
IUPAC Name dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
SMILES [O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+]

Specifications

Color Orange
CAS 7778-50-9, 7732-18-5
CAS Min % 97.0, 0.95
CAS Max % 100.0, 0.99
Synonym potassium dichromate, potassium bichromate, kaliumdichromat, iopezite, dipotassium bichromate, dipotassium dichromate, potassium dichromate vi, dichromic acid dipotassium salt, dipotassium dichromium heptaoxide, bichromate of potash
Molecular Weight (g/mol) 294.182
Grade Laboratory
Packaging Natural Poly Bottle
pH ∼4
Quantity 500 mL
Physical Form Liquid
Concentration or Composition Notes 0.200Normal
Chemical Name or Material Potassium Dichromate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.