Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Dichromate, 0.500 N (N/2), Ricca Chemical
SDP

Catalog No. 60741 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60741 4 L Natural Poly Bottle
6074-16 500 mL Natural Poly Bottle
6074-32 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 60741 Supplier Ricca Chemical Company Supplier No. 60741
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 7778-50-9 , 7732-18-5
Molecular Formula Cr2K2O7
Molecular Weight (g/mol) 294.182
InChI Key KMUONIBRACKNSN-UHFFFAOYSA-N
Synonym potassium dichromate, potassium bichromate, kaliumdichromat, iopezite, dipotassium bichromate, dipotassium dichromate, potassium dichromate vi, dichromic acid dipotassium salt, dipotassium dichromium heptaoxide, bichromate of potash
PubChem CID 24502
ChEBI CHEBI:53444
IUPAC Name dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
SMILES [O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+]

Specifications

Color Orange
CAS Min % 96.0, 2.35
CAS Max % 100.0, 2.44
Grade Laboratory
Packaging Natural Poly Bottle
pH ∼3.5 to 4
Quantity 4 L
Physical Form Liquid
Concentration or Composition Notes 0.500Normal
Chemical Name or Material Potassium Dichromate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.