Learn More
Potassium dichromate, 99%
Description
Potassium dichromate is most often used as an oxidizing agent in laboratories and industries. It is used as a precursor to prepare potassium chrome alum which finds application in leather finishing and tanning. It is utilized in the preparation of chromic acid for cleaning glassware and etching materials. It is an active ingredient in cement used for retards setting and improves its density and texture. Furthermore, it is used in photography and in photographic screen printing. It is also used in wet test analysis as an analytical lab reagent and in ethanol determination.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Potassium dichromate is most often used as an oxidizing agent in laboratories and industries. It is used as a precursor to prepare potassium chrome alum which finds application in leather finishing and tanning. It is utilized in the preparation of chromic acid for cleaning glassware and etching materials. It is an active ingredient in cement used for retards setting and improves its density and texture. Furthermore, it is used in photography and in photographic screen printing. It is also used in wet test analysis as an analytical lab reagent and in ethanol determination.
Solubility
Soluble in water. Insoluble in alcohol.
Notes
Incompatible with strong oxidizing agents, reducing agents, strong bases, organic materials, acids, powdered metals, acid anhydrides and hydrazine.
Specifications
Specifications
| Melting Point | 398°C |
| Quantity | 2500 g |
| Linear Formula | K2Cr2O7 |
| UN Number | UN3087 |
| Merck Index | 14,7627 |
| Solubility Information | Soluble in water. Insoluble in alcohol. |
| Formula Weight | 294.18 |
| Percent Purity | 99% |
| Odor | Odorless |
| Density | 2.676 |
| Show More |
Safety and Handling
| Hazard Category | H272-H301-H310+H330-H314-H317-H334-H335-H340-H350-H360FD-H372 |
| Hazard Statement | GHS H Statement H272-H301-H330-H334-H340-H350-H360-H372-H314-H312-H317 May intensify fire; oxidizer. Toxic if swallowed. Fatal if inhaled. May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause genetic defects. May cause cancer. May damage fertility or the unborn child. Causes damage to organs through prolonged or repeated exposure. Causes severe skin burns and eye damage. Harmful in contact with skin. May cause an allergic skin reaction. |
| Precautionary Statement | P201-P202-P210-P220-P221-P260-P262-P264b-P270-P271-P272-P280g-P281-P284-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, TOXIC, N.O.S. |
| EINECSNumber | 231-906-6 |
| RTECSNumber | HX7680000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.