Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium dichromate, ACS, 99.0% min

Catalog No. p-4407730
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1345022 100 g
AA1345036 500 g
AA13450A4 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. AA1345022 Supplier Thermo Scientific Chemicals Supplier No. 01345022
Only null left
Add to Cart
This item is not returnable. View return policy

For Use As An Oxidizing Agent, in Dyeing, Painting | CAS: 7778-50-9 | Cr2K2O7 | 294.182 g/mol

As an oxidizing agent, in dyeing, painting, decorating porcelain, lithography, in electric batteries, electroplating Potassium dichromate is most often used as an oxidizing agent in laboratories and industries. It is used as a precursor to prepare potassium chrome alum which finds application in leather finishing and tanning. It is utilized in the preparation of chromic acid for cleaning glassware and etching materials It is an active ingredient in cement and is used for retards setting and improves its density and texture. Furthermore, it is used in photography and in photographic screen printing. It is also used in wet test analysis as an analytical lab reagent and in ethanol determination.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Maximum level of impurities: Insoluble matter 0.005%, Loss on drying 0.05%, Cl 0.001%, SO4 0.005%, Ca 0.003%, Fe 0.001%, Na 0.02%
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7778-50-9
Molecular Formula Cr2K2O7
Molecular Weight (g/mol) 294.182
MDL Number MFCD00011367
InChI Key KMUONIBRACKNSN-UHFFFAOYSA-N
Synonym potassium dichromate, potassium bichromate, kaliumdichromat, iopezite, dipotassium bichromate, dipotassium dichromate, potassium dichromate vi, dichromic acid dipotassium salt, dipotassium dichromium heptaoxide, bichromate of potash
PubChem CID 24502
ChEBI CHEBI:53444
IUPAC Name dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
SMILES [O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+]

Specifications

Melting Point 398°C
Color Orange
Physical Form Crystalline
Quantity 100 g
Assay Percent Range 99.0% min.
Solubility Information Soluble in water. Insoluble in alcohol.
Formula Weight 294.18
Grade ACS
Appearance Orange crystalline powder
Density 2.676 g/cm3
Chemical Name or Material Potassium dichromate, For ACS analysis
Show More Show Less
Hazard Category H272-H301-H310+H330-H314-H317-H334-H335-H340-H350-H360FD-H372
Precautionary Statement P201-P202-P210-P220-P221-P260-P262-P264b-P270-P271-P272-P280g-P281-P284-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 5.1, Packing Group: II
EINECSNumber 231-906-6
RTECSNumber HX7680000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.