Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Dichromate, ACS, Reagents

Catalog No. 01337850
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-850 500 g
01-337-852 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-337-850 Supplier Reagents Holdings Llc Supplier No. C1282500500A5
Only null left
Add to Cart
Add to Cart

Potassium Dichromate, ACS, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7778-50-9
Molecular Formula Cr2K2O7
Molecular Weight (g/mol) 294.18
InChI Key KMUONIBRACKNSN-UHFFFAOYSA-N
Synonym Potassium bichromate
IUPAC Name dipotassium [(oxidodioxochromio)oxy]chromiumoylolate
SMILES [K+].[K+].[O-][Cr](=O)(=O)O[Cr]([O-])(=O)=O

Specifications

Boiling Point 500°C
Melting Point 398°C
Color Orange/Red
Physical Form Solid
Quantity 500 g
Grade ACS
Packaging Poly Bottle
Specific Gravity 2.676
Chemical Name or Material Potassium Dichromate, ACS
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.