Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Dichromate, ACS Reagent Grade, Ricca Chemical
SDP

Catalog No. RDCP042050F Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 g
12 kg
50 kg
Packaging:
Fiber Drum
White Poly Wide Mouth Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
RDCP042050F 50 kg Fiber Drum
RDCP0420500 500 g White Poly Wide Mouth Bottle
RDCP042012F 12 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. RDCP042050F Supplier Ricca Chemical Company Supplier No. RDCP042050F4
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 7778-50-9
Molecular Formula Cr2K2O7
Molecular Weight (g/mol) 294.182
InChI Key KMUONIBRACKNSN-UHFFFAOYSA-N
Synonym potassium dichromate, potassium bichromate, kaliumdichromat, iopezite, dipotassium bichromate, dipotassium dichromate, potassium dichromate vi, dichromic acid dipotassium salt, dipotassium dichromium heptaoxide, bichromate of potash
PubChem CID 24502
ChEBI CHEBI:53444
IUPAC Name dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
SMILES [O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+]

Specifications

Color Orange-Red
CAS Min % 98.0
CAS Max % 100.0
Physical Form Powder
Quantity 50 kg
Percent Purity ≥ 99%
Grade ACS
Packaging Fiber Drum
Chemical Name or Material Potassium Dichromate
ShelfLife 60 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.