Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Dichromate, Crystal, Reagent Special, ACS, 99.95-105%, Spectrum™ Chemical
SDP

Catalog No. 18605536 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-605-536 25 g Poly Bottle
18-605-537 125 g Poly Bottle
18-605-538 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 18-605-536 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P128125GM
Add to Cart
Add to Cart
This item is not returnable. View return policy

P1281, 7778-50-9, K2Cr2O7

Spectrum™ Chemical Potassium Dichromate, Crystal, Reagent Special, ACS is an inorganic chemical reagent that is most often used as an oxidizing agent in industrial applications. It is known to be chronically detrimental to health. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 7778-50-9
Molecular Formula Cr2K2O7
Molecular Weight (g/mol) 294.18
InChI Key KMUONIBRACKNSN-UHFFFAOYSA-N
IUPAC Name dipotassium [(oxidodioxochromio)oxy]chromiumoylolate
SMILES [K+].[K+].[O-][Cr](=O)(=O)O[Cr]([O-])(=O)=O

Specifications

CAS Max % 1
Quantity 25 g
Percent Purity 99.95 to 100.05%
Grade ACS
Loss on Drying 0.05
Packaging Poly Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.