Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Dichromate, (Crystalline),MP Biomedicals

Catalog No. ICN15633890 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15633890 500 g
ICN15633891 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15633890 Supplier MP Biomedicals, Inc Supplier No. 0215633890
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7778-50-9
Molecular Formula Cr2K2O7
Molecular Weight (g/mol) 294.182
InChI Key KMUONIBRACKNSN-UHFFFAOYSA-N
Synonym potassium dichromate, potassium bichromate, kaliumdichromat, iopezite, dipotassium bichromate, dipotassium dichromate, potassium dichromate vi, dichromic acid dipotassium salt, dipotassium dichromium heptaoxide, bichromate of potash
PubChem CID 24502
ChEBI CHEBI:53444
IUPAC Name dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
SMILES [O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+]

Specifications

Physical Form Crystalline
Quantity 500 g
Formula Weight 294.2
Chemical Name or Material Potassium Dichromate
Recommended Storage Room Temperature
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.